Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC13 Antibody (1280B), Biotin, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB90551B
Description
SEC13 Monoclonal specifically detects SEC13 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SEC13 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SEC13 | |
rhSEC13 aa123-322 | |
100 μL | |
metabolism | |
6396 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
1280B | |
Western Blot | |
D3S1231ESEC13L1npp-20, protein SEC13 homolog, SEC13 (S. cerevisiae)-like 1, SEC13 homolog (S. cerevisiae), SEC13L1, SEC13-like 1 isoform, SEC13-like protein 1, SEC13-related protein, SEC13RSEC13-like 1 (S. cerevisiae) | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction