Learn More
Invitrogen™ SEC13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588174
Description
Immunogen sequence: MVSVINTVDT SHEDMIHDAQ MDYYGTRLAT CSSDRSVKIF DVRNGGQILI ADLRGHEGPV WQVAWAHPMY GNILASCSYD RKVIIWREEN GTWEKSHEHA GHDSSVNSVC WAPHDYGLIL ACGSSDGAIS LLTYTGEGQW EVKKINNAHT IGCNAVSWAP AVVPGSLIDH PSGQKPNYIK RFASGGCDNL IKLWKEEEDG QWKEEQKLEA HSDWVRDVAW APSIGLPTST IASCSQDGRV FIWTCDDASS NTWSPKLLHK FNDVVWHVSW SITANILAVS GGDNKVTLWK ESVDGQWVCI SDVNKGQGSV SASVTEGQQN EQ; Positive Samples: U-87MG, HeLa, 293T, HT-29, Mouse kidney, Mouse liver, Rat ovary; Cellular Location: COPII-coated vesicle membrane, Cytoplasmic side, Cytoplasmic vesicle, Endoplasmic reticulum membrane, Nucleus, Peripheral membrane protein, nuclear pore complex.
The protein encoded by this gene belongs to the SEC13 family of WD-repeat proteins. It is a constituent of the endoplasmic reticulum and the nuclear pore complex. It has similarity to the yeast SEC13 protein, which is required for vesicle biogenesis from endoplasmic reticulum during the transport of proteins. Multiple alternatively spliced transcript variants have been found.
Specifications
| SEC13 | |
| Polyclonal | |
| Unconjugated | |
| SEC13 | |
| 1110003H02Rik; D3S1231E; GATOR complex protein SEC13; npp-20; Protein SEC13 homolog; Sec13; SEC13 homolog; SEC13 homolog (S. cerevisiae); SEC13 homolog, nuclear pore and COPII coat complex component; SEC13 homolog, nuclear pore and COPII coating complex component; SEC13 related; SEC13A; Sec13l1; SEC13-like 1; SEC13-like 1 isoform; SEC13-like protein 1; SEC13R; SEC13-related protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 110379, 297522, 6396 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P55735, Q5XFW8, Q9D1M0 | |
| SEC13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-322 of human SEC13 (NP_8991951). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.