Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC14L1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297656MFV450
Description
SEC14L1 Polyclonal antibody specifically detects SEC14L1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
SEC14L1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
DKFZp686C06176, PRELID4A, SEC14L, SEC14-like 1 (S. cerevisiae), SEC14-like protein 1 | |
Produced in rabbits immunized with E. coli-derived Human SEC14L1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
6397 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction