Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC14L1 Antibody (OTI3A6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274046H
Description
SEC14L1 Monoclonal specifically detects SEC14L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SEC14L1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SEC14L1 | |
Full length recombinant protein of human SEC14L1 (NP_002994) produced in HEK293T cells. | |
0.1 mL | |
6397.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3A6 | |
Western Blot, Immunohistochemistry | |
DKFZp686C06176, PRELID4A, SEC14L, SEC14-like 1 (S. cerevisiae), SEC14-like protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction