Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC14L1 Monoclonal Antibody (OTI3A6)
GREENER_CHOICE

Catalog No. PIMA527082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527082 Supplier Invitrogen™ Supplier No. MA527082
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SEC14L1 Monoclonal Antibody for Western Blot, IHC (P)

SEC14L1 may play a role in innate immunity by inhibiting the antiviral RIG-I signaling pathway. In this pathway, functions as a negative regulator of DDX58/RIG-I, the cytoplasmic sensor of viral nucleic acids. Prevents the interaction of DDX58 with MAVS/IPS1, an important step in signal propagation. May also regulate the SLC18A3 and SLC5A7 cholinergic transporters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC14L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SEC14L1
Gene Accession No. Q92503
Gene Alias PRELID4A; SEC14 like lipid binding 1; SEC14L; SEC14L1; SEC14-like protein 1
Gene Symbols SEC14L1
Host Species Mouse
Immunogen Full length human recombinant protein of SEC14L1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6397
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.