Learn More
Invitrogen™ SEC14L1 Polyclonal Antibody

Description
SEC14L1 Polyclonal Antibody for Western Blot, IP
Specifications
Specifications
| Antigen | SEC14L1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.03% ProClin 300; pH 7 |
| Gene | SEC14L1 |
| Gene Accession No. | Q92503 |
| Gene Alias | PRELID4A; SEC14 like lipid binding 1; SEC14L; SEC14L1; SEC14-like protein 1 |
| Gene Symbols | SEC14L1 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.