Learn More
Invitrogen™ SEC14L2 Monoclonal Antibody (OTI1F10)

Description
SEC14L2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | SEC14L2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1F10 |
| Concentration | 0.71 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SEC14L2 |
| Gene Accession No. | O76054 |
| Gene Alias | 1300013M05Rik; Alpha-tocopherol-associated protein; C22orf6; KIAA1186; KIAA1658; SEC14 like lipid binding 2; SEC14L2; SEC14-like 2; SEC14-like 2 (S. cerevisiae); SEC14-like lipid binding 2; SEC14-like protein 2; SEC14-like protein 2-like; SPF; squalene transfer protein; Supernatant protein factor; TAP; TAP1; tocopherol-associated protein; tocopherol-associated protein 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.