Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC14L2 Monoclonal Antibody (OTI1H3), TrueMAB™, OriGene
SDP

Catalog No. 502540840
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0840 30 μL
50-167-4422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0840 Supplier OriGene Supplier No. TA503561S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SEC14L2 Monoclonal antibody specifically detects SEC14L2 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen SEC14L2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1H3
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ELK3
Gene Accession No. P41970
Gene Alias 1300013M05Rik; Alpha-tocopherol-associated protein; C22orf6; KIAA1186; KIAA1658; SEC14 like lipid binding 2; SEC14L2; SEC14-like 2; SEC14-like 2 (S. cerevisiae); SEC14-like lipid binding 2; SEC14-like protein 2; SEC14-like protein 2-like; SPF; squalene transfer protein; Supernatant protein factor; TAP; TAP1; tocopherol-associated protein; tocopherol-associated protein 1
Host Species Mouse
Immunogen Full length human recombinant protein of human ELK3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2004
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.