Learn More
SEC14L2 Mouse anti-Human, Clone: OTI1H3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503561

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a cytosolic protein which belongs to a family of lipid-binding proteins including Sec14p, alpha-tocopherol transfer protein, and cellular retinol-binding protein. The encoded protein stimulates squalene monooxygenase which is a downstream enzyme in the cholesterol biosynthetic pathway. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
SEC14L2 | |
Monoclonal | |
Unconjugated | |
SEC14L2 | |
C22orf6, SPF, TAP, TAP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23541 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O76054 | |
SEC14L2 | |
Full length human recombit protein of human SEC14L2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.