Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC14L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588002 Supplier Invitrogen™ Supplier No. PA588002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRKVETITII YDCEGLGLKH LWKPAVEAYG EFLCMFEENY PETLKRLFVV KAPKLFPVAY NLIKPFLSED TRKKIMVLGA NWKEVLLKHI SPDQVPVEYG GTMTDPDGNP KCKSKINYGG DIPRKYYVRD QVKQQYEHSV QISRGSSHQV EYEILFPGCV LRWQFMSDGA DVGFGIFLKT KMGERQRAGE MTEVLPNQRY NSHLVPEDGT L; Positive Samples: Mouse brain, Mouse liver, Mouse lung, Rat liver; Cellular Location: Cytoplasm, Nucleus.

This gene encodes a cytosolic protein which belongs to a family of lipid-binding proteins including Sec14p, alpha-tocopherol transfer protein, and cellular retinol-binding protein. The encoded protein stimulates squalene monooxygenase which is a downstream enzyme in the cholesterol biosynthetic pathway. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC14L2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SEC14L2
Gene Accession No. O76054, Q99J08, Q99MS0
Gene Alias 1300013M05Rik; Alpha-tocopherol-associated protein; C22orf6; KIAA1186; KIAA1658; SEC14 like lipid binding 2; SEC14L2; SEC14-like 2; SEC14-like 2 (S. cerevisiae); SEC14-like lipid binding 2; SEC14-like protein 2; SEC14-like protein 2-like; SPF; squalene transfer protein; Supernatant protein factor; TAP; TAP1; tocopherol-associated protein; tocopherol-associated protein 1
Gene Symbols SEC14L2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-270 of human SEC14L2 (NP_0011911331).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116486, 23541, 67815
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.