Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC14L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113471
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113471 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113471 Supplier Invitrogen™ Supplier No. PA5113471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEC14L3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The protein encoded by this gene is highly similar to the protein encoded by the Saccharomyces cerevisiae SEC14 gene. The SEC14 protein is a phophatidylinositol transfer protein that is essential for biogenesis of Golgi-derived transport vesicles, and thus is required for the export of yeast secretory proteins from the Golgi complex. The specific function of this protein has not yet been determined. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC14L3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SEC14L3
Gene Accession No. Q9UDX4
Gene Alias 1110069O07Rik; 45 kDa secretory protein; rsec45; SEC14 like lipid binding 3; Sec14l3; SEC14-like 3; SEC14-like 3 (S. cerevisiae); SEC14-like lipid binding 3; SEC14-like protein 3; SEC14p-like protein TAP2; secretory protein, 45kD; Spf2; TAP2; Tocopherol-associated protein 2
Gene Symbols SEC14L3
Host Species Rabbit
Immunogen Recombinant Human SEC14-like protein 3 protein (201-400AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266629, 380683
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.