Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEC16A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144561
Description
SEC16A Polyclonal Antibody for Western Blot, ELISA
DNA methylation, or the addition of methyl groups to cytosine bases in the dinucleotide CpG, is imperative to proper development and regulates gene expression. The methylation pattern involves the enzymatic processes of methylation and demethylation. The demethylation enzyme was recently found to be a mammalian protein, which exhibits demethylase activity associated to a methyl-CpG-binding domain (MBD). The enzyme is able to revert methylated cytosine bases to cytosines within the particular dinucleotide sequence mdCpdG by catalyzing the cleaving of the methyl group as methanol. MeCP2 and MBD1 (PCM1) are first found to repress transcription by binding specifically to methylated DNA. MBD2 and MBD4 (also known as MED1) were later found to colocalize with foci of heavily methylated satellite DNA and believed to mediate the biological functions of the methylation signal. Surprisingly, MBD3 does not bind methylated DNA both in vivo and in vitro. MBD1, MBD2, MBD3, and MBD4 are found to be expressed in somatic tissues, but the expression of MBD1 and MBD2 is reduced or absent in embryonic stem cells, which are known to be deficient in MeCP1 activity. MBD4 have homology to bacterial base excision repair DNA N-glycosylases/lyases. In some microsatellite unstable tumors MBD4 is mutated at an exonic polynucleotide tract.
Specifications
SEC16A | |
Polyclonal | |
Unconjugated | |
SEC16 | |
KIAA0310; p250; protein SEC16 homolog A; protein transport protein Sec16A; SEC16; SEC16 homolog A; SEC16 homolog A, endoplasmic reticulum export factor; SEC16A; SEC16L | |
Rabbit | |
Affinity chromatography | |
RUO | |
856020 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P48415 | |
SEC16 | |
Recombinant Saccharomyces cerevisiae (strain ATCC 204508/S288c) (Baker's yeast) SEC16 protein (280-430aa). | |
100 μL | |
Primary | |
Yeast | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction