Learn More
SEC16A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SEC16A |
---|---|
Concentration | 240 μg/mL |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8409
![]() ![]() |
Proteintech Group Inc
200251AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The SEC16A antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human SEC16A. This antibody recognizes human, mouse, rat antigen. The SEC16A antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
SEC16A | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
240 μg/mL | |
Polyclonal | |
Rabbit | |
9919 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.