Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC22A Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144171
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144171 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144171 Supplier Invitrogen™ Supplier No. PA5144171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEC22A Polyclonal Antibody for ICC/IF, IHC (P), ELISA

The protein encoded by this gene belongs to the member of the SEC22 family of vesicle trafficking proteins. This protein has similarity to rat SEC22 and may act in the early stages of the secretory pathway. [provided by RefSeq, Nov 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC22A
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene SEC22A
Gene Accession No. Q96IW7
Gene Alias SEC22 vesicle-trafficking protein homolog A; SEC22 vesicle-trafficking protein-like 2; SEC22A; SEC22L2; Vesicle-trafficking protein SEC22a
Gene Symbols SEC22A
Host Species Rabbit
Immunogen Recombinant Human Vesicle-trafficking protein SEC22a protein (10-187AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26984
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.