Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC22C Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200034 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018502 0.1 mg
NBP130760 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018502 Supplier Novus Biologicals Supplier No. NBP1307600.1MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEC22C Polyclonal specifically detects SEC22C in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen SEC22C
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Sterilized water with No Preservative
Gene Alias DKFZp761F2321, MGC13261, MGC5373, SEC22 vesicle trafficking protein homolog C (S. cerevisiae), SEC22 vesicle trafficking protein-like 3, SEC22 vesicle trafficking protein-like 3 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog C, SEC22 vesicle-trafficking protein-like 3, SEC22L3, secretion deficient 22C, vesicle-trafficking protein SEC22c
Gene Symbols SEC22C
Host Species Rabbit
Immunogen A synthetic from the c-terminal of human Vesicle trafficking protein SEC22C (SEC22L3) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in mouse and shares 95% identity with rat sequence.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9117
Test Specificity Appears to be specific for SEC22C.
Reconstitution Reconstitute with 0.5 ml sterilized water.
Target Species Mouse, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.