Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC22C Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SEC22C |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SEC22C Polyclonal specifically detects SEC22C in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SEC22C | |
Polyclonal | |
Rabbit | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
9117 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TASYDTTCIGLASRPYAFLEFDSIIQKVKWHFNYVSSSQMECSLEKIQEELKLQPPAVLTLEDTDVANGVMNGHTPMHL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
DKFZp761F2321, MGC13261, MGC5373, SEC22 vesicle trafficking protein homolog C (S. cerevisiae), SEC22 vesicle trafficking protein-like 3, SEC22 vesicle trafficking protein-like 3 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog C, SEC22 vesicle-trafficking protein-like 3, SEC22L3, secretion deficient 22C, vesicle-trafficking protein SEC22c | |
SEC22C | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title