Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC23A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588348 Supplier Invitrogen™ Supplier No. PA588348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTTYLEFIQQ NEERDGVRFS WNVWPSSRLE ATRMVVPVAA LFTPLKERPD LPPIQYEPVL CSRTTCRAVL NPLCQVDYRA KLWACNFCYQ RNQFPPSYAG ISELNQPAEL LPQFSSIEYV VLRGPQMPLI FLYVVDTCME DEDLQALKES; Positive Samples: B-cell, LO2, HeLa, SGC-7901, HT-29, Mouse brain, Mouse liver, Rat brain; Cellular Location: Golgi apparatus membrane, Peripheral membrane protein, Smooth endoplasmic reticulum membrane.

The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family. It is part of a protein complex and found in the ribosome-free transitional face of the endoplasmic reticulum (ER) and associated vesicles. This protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The encoded protein is suggested to play a role in the ER-Golgi protein trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC23A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Sec23a
Gene Accession No. Q01405, Q15436
Gene Alias CLSD; hSec23A; MGC26267; Msec23; protein transport protein Sec23A; Sec23; Sec23 homolog A (S. cerevisiae); Sec23 homolog A, coat complex II component; Sec23 homolog A, COPII coat complex component; Sec23a; SEC23A (S. cerevisiae); Sec23r; SEC23-related protein A
Gene Symbols Sec23a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human Sec23A (NP_0063552).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10484, 20334, 58817
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.