Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEC23A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144934
Description
Immunogen sequence: DLSAKQLQEM LGLSKVPLTQ ATRGPQVQQP PPSNRFLQPV Q Antigen sequence identity: Mouse 98%; Rat 98%.
The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family. It is part of a protein complex and found in the ribosome-free transitional face of the endoplasmic reticulum (ER) and associated vesicles. This protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The encoded protein is suggested to play a role in the ER-Golgi protein trafficking.
Specifications
SEC23A | |
Polyclonal | |
Unconjugated | |
Sec23a | |
CLSD; hSec23A; MGC26267; Msec23; protein transport protein Sec23A; Sec23; Sec23 homolog A (S. cerevisiae); Sec23 homolog A, coat complex II component; Sec23 homolog A, COPII coat complex component; Sec23a; SEC23A (S. cerevisiae); Sec23r; SEC23-related protein A | |
Rabbit | |
Affinity chromatography | |
RUO | |
10484 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15436 | |
Sec23a | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 194-234. | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction