Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sec23a Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519011
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519011 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519011 Supplier Invitrogen™ Supplier No. PA519011
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, equine, human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family. It is part of a protein complex and found in the ribosome-free transitional face of the endoplasmic reticulum (ER) and associated vesicles. This protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The encoded protein is suggested to play a role in the ER-Golgi protein trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sec23a
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Sec23a
Gene Accession No. Q01405
Gene Alias CLSD; hSec23A; MGC26267; Msec23; protein transport protein Sec23A; Sec23; Sec23 homolog A (S. cerevisiae); Sec23 homolog A, coat complex II component; Sec23 homolog A, COPII coat complex component; Sec23a; SEC23A (S. cerevisiae); Sec23r; SEC23-related protein A
Gene Symbols Sec23a
Host Species Goat
Immunogen Synthetic peptide sequence (ANRAATTGHVID) corresponding to the internal amino acids of SEC23A (aa 335-347).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20334
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.