Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC23A Recombinant Rabbit Monoclonal Antibody (JU37-48)
GREENER_CHOICE

Catalog No. PIMA534630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534630 Supplier Invitrogen™ Supplier No. MA534630
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: NIH-3T3, HepG2, HUVEC, human tonsil tissue, human fetal skeletal muscle tissue, HepG2.

The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family. It is part of a protein complex and found in the ribosome-free transitional face of the endoplasmic reticulum (ER) and associated vesicles. This protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The encoded protein is suggested to play a role in the ER-Golgi protein trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC23A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JU37-48
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Sec23a
Gene Accession No. Q15436
Gene Alias CLSD; hSec23A; MGC26267; Msec23; protein transport protein Sec23A; Sec23; Sec23 homolog A (S. cerevisiae); Sec23 homolog A, coat complex II component; Sec23 homolog A, COPII coat complex component; Sec23a; SEC23A (S. cerevisiae); Sec23r; SEC23-related protein A
Gene Symbols Sec23a
Host Species Rabbit
Immunogen Recombinant protein within Human SEC23 aa 280-420.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10484
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.