Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC23B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156339
Description
SEC23B Polyclonal antibody specifically detects SEC23B in Human, Mouse samples. It is validated for Western Blot.Specifications
SEC23B | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CDAII, CDA-II, CDAN2, congenital dyserythropoietic anemia, type II, HEMPASSec23 (S. cerevisiae) homolog B, protein transport protein Sec23B, Sec23 homolog B (S. cerevisiae), SEC23-like protein B, SEC23-related protein B, transport protein SEC23B | |
Rabbit | |
Affinity purified | |
RUO | |
10483 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q15437 | |
SEC23B | |
Synthetic peptides corresponding to SEC23B(Sec23 homolog B (S. cerevisiae)) The peptide sequence was selected from the middle region of SEC23B. Peptide sequence SFSLYPQFMFHLRRSPFLQVFNNSPDESSYYRHHFARQDLTQSLIMIQPI. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Xenopus: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction