Learn More
SEC23B Mouse anti-Human, Clone: OTI8G4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810159

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The function of this gene product has been implicated in cargo selection and concentration. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
SEC23B | |
Monoclonal | |
Unconjugated | |
SEC23B | |
CDA-II, CDAII, CDAN2, CWS7, HEMPAS, hSec23B | |
Mouse | |
Affinity Chromatography | |
RUO | |
10483 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI8G4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15437 | |
SEC23B | |
Human recombit protein fragment corresponding to amino acids 392-641 of human SEC23B produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.