Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC23B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584830 Supplier Invitrogen™ Supplier No. PA584830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IDIYACALDQ TGLLEMKCCA NLTGGYMVMG DSFNTSLFKQ TFQRIFTKD Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.

The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The function of this gene product has been implicated in cargo selection and concentration. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC23B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEC23B
Gene Accession No. Q15437
Gene Alias CDAII; CDA-II; CDAN2; CWS7; HEMPAS; hSec23B; Protein transport protein Sec23B; Sec23 homolog B; Sec23 homolog B (S. cerevisiae); Sec23 homolog B, coat complex II component; Sec23 homolog B, COPII coat complex component; Sec23b; SEC23B (S. cerevisiae); SEC23-like protein B; SEC23-related protein B; transport protein SEC23B
Gene Symbols SEC23B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEC23B. Recombinant protein control fragment (Product #RP-105494).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10483
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.