Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC23IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1001561
Description
SEC23IP Polyclonal specifically detects SEC23IP in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SEC23IP | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
P125, P125A, SEC23 interacting protein, SEC23-interacting protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2500-1:10000, Immunoprecipitation 1-4 ug/mg of lysate | |
NP_009121.1 | |
SEC23IP | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Sec23-Interacting Protein using the numbering given in entry NP_009121.1 (GeneID 11196). | |
0.1 mL | |
Golgi Apparatus Markers | |
11196 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction