Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC23IP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543797 Supplier Invitrogen™ Supplier No. PA543797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VPFIPVTQAS ASPASLLLPG EDSTDVGEED SFLGQTSIHT SAPQTFSYFS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.

SEC23IP is involved in protein transport from the endoplasmic reticulum to the Golgi apparatus. The protein encoded by this gene was identified by its interaction with a mouse protein similar to yeast Sec23p, an essential component of the COPII. This protein shares significant similarity with phospholipid-modifying proteins, especially phosphatidic acid preferring-phospholipase A1. Overexpression of this protein has been shown to cause disorganization of the endoplasmic reticulum-Golgi intermediate compartment and Golgi apparatus, which suggests its role in the early secretory pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC23IP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SEC23IP
Gene Accession No. Q9Y6Y8
Gene Alias D7Ertd373e; MSTP053; P125; P125A; SEC23 interacting protein; SEC23-interacting protein; Sec23-interacting protein p125; SEC23-interacting protein-like protein; SEC23IP
Gene Symbols SEC23IP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SEC23IP (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11196
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.