Learn More
Invitrogen™ SEC24A Polyclonal Antibody

Description
Antibody detects endogenous levels of total SEC24A.
Specifications
Specifications
| Antigen | SEC24A |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SEC24A |
| Gene Accession No. | O95486, Q3U2P1 |
| Gene Alias | Protein transport protein Sec24A; SEC24 family member A; SEC24 family, member A; SEC24 homolog A, COPII coat complex component; SEC24 related gene family, member A; Sec24 related gene family, member A (S. cerevisiae); Sec24a; SEC24-related protein A |
| Gene Symbols | SEC24A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.