Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC24A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566043 Supplier Invitrogen™ Supplier No. PA566043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NGPVQNALLSS QESVSQGYNF QLPGSYPHPI PAKTLNPVSG QSNYGGSQGS GQTLNRPPVA SNPVTPSLHS GPAPRMPLPA SQNPATTPMP Highest antigen sequence identity to the following orthologs - mouse 77%, rat 78%.

SC24A is a component of coat protein II (COPII; see MIM 601924)-coated vesicles that mediate protein transport from the endoplasmic reticulum. SEC24A is 1 of several mammalian proteins that show structural and functional homology to yeast Sec24.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC24A
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEC24A
Gene Accession No. O95486
Gene Alias Protein transport protein Sec24A; SEC24 family member A; SEC24 family, member A; SEC24 homolog A, COPII coat complex component; SEC24 related gene family, member A; Sec24 related gene family, member A (S. cerevisiae); Sec24a; SEC24-related protein A
Gene Symbols SEC24A
Host Species Rabbit
Immunogen Recombinant Human SEC24A. Recombinant protein control fragment (Product #RP-105055).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.