Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC24B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558284 Supplier Invitrogen™ Supplier No. PA558284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFQGAASSAS HLHTSASQPY SSFVNHYNSP AMYSASSSVA SQGFPSTCGH YAMSTVSNAA YPSVSYPSLP AGDTYGQMFT SQNAPTVR Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 69%.

Component of the coat protein complex II (COPII) which promotes the formation of transport vesicles from the endoplasmic reticulum (ER). The coat has two main functions, the physical deformation of the endoplasmic reticulum membrane into vesicles and the selection of cargo molecules for their transport to the Golgi complex. Plays a central role in cargo selection within the COPII complex and together with SEC24A may have a different specificity compared to SEC24C and SEC24D. May package preferentially cargos with cytoplasmic DxE or LxxLE motifs and may also recognize conformational epitopes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC24B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEC24B
Gene Accession No. O95487
Gene Alias AI605202; Protein transport protein Sec24B; SEC24; SEC24 homolog B, COPII coat complex component; SEC24 related gene family, member B; Sec24 related gene family, member B (S. cerevisiae); Sec24b; SEC24-related protein B; secretory protein 24
Gene Symbols SEC24B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEC24B. Recombinant protein control fragment (Product #RP-96669).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10427
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.