Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC24C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110348 Supplier Invitrogen™ Supplier No. PA5110348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVLQPGAEVT TDDRAYVRQL VTSMDVTETN VFFYPRLLPL TKSPVESTTE PPAVRASEER LSNGDIYLLE NGLNLFLWVG ASVQQGVVQS LFSVSSFSQI TSGLSVLPVL DNPLSKKVRG LIDSLRAQRS RYMKLTVVKQ EDKMEMLFKH FLVEDKSLSG GASYVDFLCH MHKEIRQLLS.

Interacts with HSP70 and can stimulate its ATPase activity. Stimulates the association between HSC70 and HIP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC24C
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SEC24C
Gene Accession No. P53992
Gene Alias 2610204K03Rik; KIAA0079; mKIAA0079; Protein transport protein Sec24C; SEC24 family member C; SEC24 family, member C; SEC24 homolog C, COPII coat complex component; SEC24 related gene family, member C; Sec24 related gene family, member C (S. cerevisiae); Sec24c; SEC24-related protein C
Gene Symbols SEC24C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 915-1094 of human SEC24C (NP_004913.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9632
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.