Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC24D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590925 Supplier Invitrogen™ Supplier No. PA590925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AASQLILPDS MKVLPVYMNC LLKNCVLLSR PEISTDERAY QRQLVMTMGV ADSQLFFYPQ LLPIHTLDVK STMLPAAVRC SESRLSEEGI FLLANGLHMF LWLGVSSPPE LIQGIFNVPS FAHINTDMTL LPEVGNPYSQ QLRMIMGIIQ QKRPYSMKLT IVKQREQPEM V; Positive Samples: LO2, BxPC-3, A-375, DU145, Mouse liver; Cellular Location: Cytoplasm, Endoplasmic reticulum membrane, Golgi apparatus membrane, perinuclear region.

The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. This gene product is implicated in the shaping of the vesicle, and also in cargo selection and concentration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC24D
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SEC24D
Gene Accession No. O94855
Gene Alias 2310020L09Rik; CLCRP2; Gm1349; KIAA0755; Protein transport protein Sec24D; SEC24 family member D; SEC24 family, member D; SEC24 homolog D, COPII coat complex component; SEC24 related gene family, member D; Sec24 related gene family, member D (S. cerevisiae); SEC24D; SEC24-related protein D
Gene Symbols SEC24D
Host Species Rabbit
Immunogen Recombinant protein (or fragment)).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310843, 69608, 9871
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.