Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC3 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178752DL594
Description
SEC3 Polyclonal antibody specifically detects SEC3 in Human, Mouse samples. It is validated for Western BlotSpecifications
SEC3 | |
Polyclonal | |
Western Blot | |
BM-102, exocyst complex component 1, Exocyst complex component Sec3, FLJ10893, SEC3L1, SEC3-like 1, SEC3-like 1 (S. cerevisiae), SEC3P, SEC3Sec3p | |
The immunogen for this product maps to a region between residue 100 and 150 of human Exocyst Complex Component Sec3 using the numbering given in entry NP_060731.2 (GeneID 55763). | |
0.1 mL | |
Signal Transduction | |
55763 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction