Learn More
Sec3 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303363A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of the exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
Sec3 | |
Polyclonal | |
Unconjugated | |
EXOC1 | |
BM-102, SEC3, SEC3L1, SEC3P, SEC3-like 1, exocyst complex component Sec3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55763, 69940 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8R3S6, Q9NV70 | |
Exoc1 | |
Between 100 and 150 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.