Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC31A Monoclonal Antibody (5D12C6)
GREENER_CHOICE

Catalog No. PIMA531900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531900 Supplier Invitrogen™ Supplier No. MA531900
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31900 has been tested in indirect ELISA.

The protein encoded by this gene is similar to yeast Sec31 protein. Yeast Sec31 protein is known to be a component of the COPII protein complex which is responsible for vesicle budding from endoplasmic reticulum (ER). This protein was found to colocalize with SEC13, one of the other components of COPII, in the subcellular structures corresponding to the vesicle transport function. An immunodepletion experiment confirmed that this protein is required for ER-Golgi transport. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC31A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5D12C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SEC31A
Gene Accession No. O94979
Gene Alias 1810024J13Rik; ABP125; ABP130; AU042160; cb354; HSPC275; HSPC334; KIAA0905; LOW QUALITY PROTEIN: protein transport protein Sec31A; protein transport protein Sec31A; SEC31 homolog A (S. cerevisiae); SEC31 homolog A, COPII coat complex component; SEC31 homolog A, COPII coating complex component; sec31a; SEC31L1; SEC31-like 1; SEC31-like protein 1; sec31p; sec31p homolog; SEC31-related protein A; Unknown (protein for MGC:63547); VAP1; vesicle associated protein; vesicle-associated protein 1; Web1-like protein; yeast Sec31p homolog; zgc:63547
Gene Symbols SEC31A
Host Species Mouse
Immunogen Purified recombinant fragment of human SEC31A (AA: 429-571) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22872
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.