Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC31A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597205
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597205 Supplier Invitrogen™ Supplier No. PA597205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTNTQWCFDI QWCPRNPAVL SAASFDGRIS VYSIMGGSTD GLRQKQVDKL SSSFGNLDPF GTGQPLPPLQ IPQQTAQHSI VLPLKKPPKW IRRPVGASFS FGGKLVTFEN VRMPSHQGAE QQQQQHHVFI SQVVTEKEFL SRSDQLQQAV QSQGFINYCQ KKIDASQTEF EKNVWSFLKV NFEDDSRGKY LELLGYRKED L; Positive Samples: Mouse liver, Rat liver; Cellular Location: COPII-coated vesicle membrane, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle, Endoplasmic reticulum membrane, Peripheral membrane protein.

The protein encoded by this gene is similar to yeast Sec31 protein. Yeast Sec31 protein is known to be a component of the COPII protein complex which is responsible for vesicle budding from endoplasmic reticulum (ER). This protein was found to colocalize with SEC13, one of the other components of COPII, in the subcellular structures corresponding to the vesicle transport function. An immunodepletion experiment confirmed that this protein is required for ER-Golgi transport. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC31A
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SEC31A
Gene Accession No. O94979, Q3UPL0, Q9Z2Q1
Gene Alias 1810024J13Rik; ABP125; ABP130; AU042160; cb354; HSPC275; HSPC334; KIAA0905; LOW QUALITY PROTEIN: protein transport protein Sec31A; protein transport protein Sec31A; SEC31 homolog A (S. cerevisiae); SEC31 homolog A, COPII coat complex component; SEC31 homolog A, COPII coating complex component; sec31a; SEC31L1; SEC31-like 1; SEC31-like protein 1; sec31p; sec31p homolog; SEC31-related protein A; Unknown (protein for MGC:63547); VAP1; vesicle associated protein; vesicle-associated protein 1; Web1-like protein; yeast Sec31p homolog; zgc:63547
Gene Symbols SEC31A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-500 of human SEC31A (NP_0557482).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22872, 69162, 93646
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.