Learn More
Invitrogen™ SEC31A Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 91%.
Specifications
Specifications
| Antigen | SEC31A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SEC31A |
| Gene Accession No. | Q9Z2Q1 |
| Gene Alias | 1810024J13Rik; ABP125; ABP130; AU042160; cb354; HSPC275; HSPC334; KIAA0905; LOW QUALITY PROTEIN: protein transport protein Sec31A; protein transport protein Sec31A; SEC31 homolog A (S. cerevisiae); SEC31 homolog A, COPII coat complex component; SEC31 homolog A, COPII coating complex component; sec31a; SEC31L1; SEC31-like 1; SEC31-like protein 1; sec31p; sec31p homolog; SEC31-related protein A; Unknown (protein for MGC:63547); VAP1; vesicle associated protein; vesicle-associated protein 1; Web1-like protein; yeast Sec31p homolog; zgc:63547 |
| Gene Symbols | SEC31A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.