Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC6 Antibody (9H5), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications
$268.00 - $743.50
Specifications
Antigen | SEC6 |
---|---|
Clone | 9H5 |
Dilution | Western Blot 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoblotting |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018792
![]() |
Novus Biologicals
NBP1975000.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP19750005
![]() |
Novus Biologicals
NBP1975000.05MG |
0.05 mg |
Each for $390.00
|
|
|||||
NBP19750002
![]() |
Novus Biologicals
NBP1975000.2MG |
0.2 mg |
Each for $743.50
|
|
|||||
Description
SEC6 Monoclonal specifically detects SEC6 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep, Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
SEC6 | |
Western Blot 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoblotting | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
11336 | |
Recombinant rat Sec6. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
9H5 | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Chicken, Mouse, Sheep, Rabbit, Pig, Monkey, Rat, Human, Canine, Bovine, Hamster | |
exocyst complex component 3, Exocyst complex component Sec6, Sec 6 homolog, SEC6, SEC6L1, SEC6-like 1, SEC6-like 1 (S. cerevisiae), Sec6p | |
EXOC3 | |
IgG2a | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title