Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC61A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521773 Supplier Invitrogen™ Supplier No. PA521773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: K562 whole cell extract, K562 membrane fraction extract (un-boiled). Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Dog (100%), Pig (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Sec61 complex is responsible for translocation of proteins across the rough endoplasmic reticulum membrane. Sec61 is composed of three subunits: alpha, beta, and gamma; that associate to form a heterotrimer. Multiple heterotrimers associate forming the ER translocon. Sec61 alpha is a polytopic protein that has ten membrane-spanning loops. Sec61 has been found to be tightly associated with membrane bound ribosomes, either directly or through adaptor proteins, and is required for assembly of membrane and secretory proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC61A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SEC61A1
Gene Accession No. P61619
Gene Alias AA408394; AA410007; homologue to sec61; HSEC61; protein transport protein SEC61 alpha subunit; protein transport protein Sec61 subunit alpha; protein transport protein Sec61 subunit alpha isoform 1; protein transport protein Sec61 subunit alpha isoform 1; protein transport protein Sec61 subunit alpha; SEC61; Sec61 alpha 1 subunit; Sec61 alpha 1 subunit (S. cerevisiae); Sec61 alpha subunit homolog; sec61 alpha-1; sec61 homolog; sec61 homologue; SEC61 translocon alpha 1 subunit; SEC61 translocon subunit alpha 1; SEC61, alpha subunit; SEC61A; SEC61A1; transport protein sec61 alpha subunit
Gene Symbols SEC61A1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SEC61A1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29927
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.