Learn More
Invitrogen™ SEC61alpha Recombinant Rabbit Monoclonal Antibody (9W7I3)

Description
Positive Samples: 293T, SH-SY5Y, Mouse lung, Mouse testis, Mouse brain, Rat testis Immunogen sequence: SMGAIFEDPV HVVVYIIFML GSCAFFSKTW IEVSGSSAKD VAKQLKEQQM VMRGHRDTSM VHELNRYIPT AAAFGGLCIG ALSVLADFLG AIGSGTGILL A.
Specifications
Specifications
| Antigen | SEC61alpha |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 9W7I3 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SEC61A2 |
| Gene Accession No. | Q9H9S3, Q9JLR1 |
| Gene Alias | protein transport protein Sec61 subunit alpha; protein transport protein Sec61 subunit alpha isoform 2; Sec61 alpha; Sec61 alpha 2 subunit; Sec61 alpha 2 subunit (S. cerevisiae); Sec61 alpha form 2; sec61 alpha-2; Sec61 translocon alpha 2 subunit; Sec61, alpha subunit 2; Sec61, alpha subunit 2 (S. cerevisiae); SEC61A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.