Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC62 Antibody (SR1865), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32262750UL
Description
SEC62 Monoclonal antibody specifically detects SEC62 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, SerumSpecifications
SEC62 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1865 | |
Western Blot 1:500-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Serum 1:50-1:200 | |
Dtrp1, hTP-1, S.cerevisiae, homolog of, SEC62 homolog (S. cerevisiae), TLOC1, TP-1, translocation protein 1, translocation protein SEC62 | |
A synthesized peptide derived from human SEC62 (Uniprot #: Q99442) | |
50 μg | |
Signal Transduction | |
7095 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction