Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEC62 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563960
Description
Immunogen sequence: MAERRRHKKR IQEVGEPSKE EKAVAKYLRF NCPTKSTNMM GHRVDYFIAS KAVDCLLDSK WAKAKK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
The Sec61 complex is the central component of the protein translocation apparatus of the endoplasmic reticulum (ER) membrane. The protein encoded by this gene and SEC63 protein are found to be associated with ribosome-free SEC61 complex. It is speculated that Sec61-Sec62-Sec63 may perform post-translational protein translocation into the ER. The Sec61-Sec62-Sec63 complex might also perform the backward transport of ER proteins that are subject to the ubiquitin-proteasome-dependent degradation pathway. The encoded protein is an integral membrane protein located in the rough ER. [provided by RefSeq].
Specifications
SEC62 | |
Polyclonal | |
Unconjugated | |
SEC62 | |
3100002M17Rik; Ab2-292; AI844545; AW545092; Dtrp1; HTP1; hTP-1; membrane protein SEC62, S.cerevisiae, homolog of; Sec62; SEC62 homolog; SEC62 homolog (S. cerevisiae); SEC62 homolog, preprotein translocation factor; SEC62 preprotein translocation factor; Tloc1; TP-1; translocation protein 1; Translocation protein SEC62 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7095 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99442 | |
SEC62 | |
Recombinant protein corresponding to Human SEC62. Recombinant protein control fragment (Product #RP-103403). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction