Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC62 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572702
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572702 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572702 Supplier Invitrogen™ Supplier No. PA572702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEC62 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The Sec61 complex is the central component of the protein translocation apparatus of the endoplasmic reticulum (ER) membrane. The protein encoded by this gene and SEC63 protein are found to be associated with ribosome-free SEC61 complex. It is speculated that Sec61-Sec62-Sec63 may perform post-translational protein translocation into the ER. The Sec61-Sec62-Sec63 complex might also perform the backward transport of ER proteins that are subject to the ubiquitin-proteasome-dependent degradation pathway. The encoded protein is an integral membrane protein located in the rough ER. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC62
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SEC62
Gene Accession No. Q8BU14, Q99442
Gene Alias 3100002M17Rik; Ab2-292; AI844545; AW545092; Dtrp1; HTP1; hTP-1; membrane protein SEC62, S.cerevisiae, homolog of; Sec62; SEC62 homolog; SEC62 homolog (S. cerevisiae); SEC62 homolog, preprotein translocation factor; SEC62 preprotein translocation factor; Tloc1; TP-1; translocation protein 1; Translocation protein SEC62
Gene Symbols SEC62
Host Species Rabbit
Immunogen SEC62 antibody for an 18 amino acid synthetic peptide near the carboxy terminus of human SEC62. The immunogen is within amino acids 270 - 320 of SEC62.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294912, 69276, 7095
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.