Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC63 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542936 Supplier Invitrogen™ Supplier No. PA542936
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: WWLYIADRKE QTLISMPYHV CTLKDTEEVE LKFPAPGKPG NYQYTVFLRS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

SEC63 gene ontology annotations related to this gene include liver development; multicellular organism aging; nitrogen compound metabolic process; posttranslational protein targeting to endoplasmic reticulum membrane; posttranslational protein targeting to membrane, translocation; renal system development; SRP-dependent cotranslational protein targeting to membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC63
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SEC63
Gene Accession No. Q9UGP8
Gene Alias 5730478J10Rik; AI649014; AW319215; DNAJC23; ERdj2; PRO2507; SEC63; SEC63 homolog; SEC63 homolog (S. cerevisiae); SEC63 homolog, protein translocation regulator; SEC63 protein translocation regulator; SEC63L; SEC63-like; SEC63-like (S. cerevisiae); SEC63-like protein; Translocation protein SEC63 homolog; translocation protein SEC63 homolog; LOW QUALITY PROTEIN: translocation protein SEC63 homolog; zgc:92718
Gene Symbols SEC63
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SEC63 (aa 651-700).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11231
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.