Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC63 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563873 Supplier Invitrogen™ Supplier No. PA563873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDEEEVSDKG SDSEEEETNR DSQSEKDDGS DRDSDREQDE KQNKDDEAEW QELQQSIQRK ERALLETKS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 94%.

SEC63 gene ontology annotations related to this gene include liver development; multicellular organism aging; nitrogen compound metabolic process; posttranslational protein targeting to endoplasmic reticulum membrane; posttranslational protein targeting to membrane, translocation; renal system development; SRP-dependent cotranslational protein targeting to membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC63
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEC63
Gene Accession No. Q9UGP8
Gene Alias 5730478J10Rik; AI649014; AW319215; DNAJC23; ERdj2; PRO2507; SEC63; SEC63 homolog; SEC63 homolog (S. cerevisiae); SEC63 homolog, protein translocation regulator; SEC63 protein translocation regulator; SEC63L; SEC63-like; SEC63-like (S. cerevisiae); SEC63-like protein; Translocation protein SEC63 homolog; translocation protein SEC63 homolog; LOW QUALITY PROTEIN: translocation protein SEC63 homolog; zgc:92718
Gene Symbols SEC63
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEC63. Recombinant protein control fragment (Product #RP-103377).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.