Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sec8 Antibody (14G1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

$203.51 - $739.91

Specifications

Antigen Sec8
Clone 14G1
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 12.5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018786
SDP
View Documents
Novus Biologicals
NBP1974920.025MG
0.025 mg
Each for $203.51
Only null left
Add to Cart
 
NBP19749205
SDP
View Documents
Novus Biologicals
NBP1974920.05MG
0.05 mg
Each for $370.21
Only null left
Add to Cart
 
NBP19749202
SDP
View Documents
Novus Biologicals
NBP1974920.2MG
0.2 mg
Each for $739.91
Only null left
Add to Cart
 
Description

Description

Sec8 Monoclonal specifically detects Sec8 in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Sec8
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 12.5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Monoclonal
Purified
RUO
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
60412
Recombinant rat rSec8.
Primary
Store at -20C. Avoid freeze-thaw cycles.
14G1
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Unconjugated
Mouse
Bovine, Canine, Fish, Hamster, Primate, Guinea Pig, Rabbit, Sheep, Mouse, Human, Rat, Chicken
exocyst complex component 4, KIAA1699SEC8L1, MGC27170, REC8, SEC8Exocyst complex component Sec8, SEC8-like 1, SEC8-like 1 (S. cerevisiae), Sec8p
EXOC4
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.