Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Secernin-1 Antibody (OTI1F2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274052B
Description
Secernin-1 Monoclonal specifically detects Secernin-1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Secernin-1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SCRN1 | |
Full length recombinant protein of human Secernin-1 (NP_055581) produced in HEK293T cells. | |
0.1 mL | |
9805.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1F2 | |
Western Blot, Immunohistochemistry | |
KIAA0193SES1, secernin 1, secernin-1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction