Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SECISBP2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222301C
Description
SECISBP2 Polyclonal antibody specifically detects SECISBP2 in Human samples. It is validated for Western BlotSpecifications
SECISBP2 | |
Polyclonal | |
Western Blot | |
DKFZp686C09169, SBP2SECIS-binding protein 2, SECIS binding protein 2, selenocysteine insertion sequence binding protein 2, selenocysteine insertion sequence-binding protein 2 | |
The immunogen this antibody was made to, maps to a region between residue 804 to 854 of human SECIS Binding Protein 2 using the numbering given in entry NP_076982.3 (GeneID 79048). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79048 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction