Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Secnidazole, Thermo Scientific™

Catalog No. p-7227423
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Type Secnidazole
Chemical Name or Material Secnidazole
Physical Form Powder
CAS 3366-95-8
Melting Point 70°C to 76°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Water ≤6 %
Color White to Yellow
Quantity 1 g
Molecular Formula C7 H11 N3 O3
Product Line Life science
Shipping Condition No special requirements
MDL Number 00864656
Merck Index XIII 8494
Solubility Information Soluble in methanol
Molecular Weight (g/mol) 185.18
Percent Purity ≥97.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.