Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Secobarbital Mouse, Unconjugated, Clone: P01-99-41M-P, Abnova™

Catalog No. 89339341 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-341 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-341 Supplier Abnova Corporation Supplier No. MAB21955
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Secobarbital Monoclonal antibody specifically detects Secobarbital in samples. It is validated for ELISA, LFIA.

Specifications

Antigen Secobarbital
Applications ELISA, Lateral Flow
Classification Monoclonal
Clone P01-99-41M-P
Conjugate Unconjugated
Host Species Mouse
Immunogen Secobarbital (conjugated with KLH) linked off the 5 position
Quantity 1 mg
Primary or Secondary Primary
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.