Learn More
Invitrogen™ Secobarbital Polyclonal Antibody

Description
Working Concentration: 1/1K. Sensitivity: 10 ng/mL Secobarbital produces 52% inhibition in a competitive ELISA employing Secobarbital polyclonal antibody.
Specifications
Specifications
| Antigen | Secobarbital |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene Alias | C12H18N2O3; Meballymal; Quinalbarbital; Quinalbarbitone; Secobarbitone; Seconal |
| Host Species | Sheep |
| Immunogen | Secobarbital(5)-BSA. |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.