Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Second Strand Buffer

Catalog No. 10812014
Encompass_Preferred
Click to view available options
Quantity:
0.5 mL

Catalog No. 10-812-014

Supplier: Invitrogen™ 10812014

Only null left
Add to Cart
Add to Cart

Includes

Supplied at a 5X concentration.

Produce second-strand cDNA from mRNA

Second Strand Buffer is used with SuperScript™ cDNA Synthesis Systems to generate second-strand cDNA from mRNA in the first-strand reaction. It is supplied as a 5X concentrate and should be diluted 1:5 (1 part 5X Second Strand Buffer and 4 parts other components) prior to use.
Quality control This product has passed quality control assays that verify the absence of detectable levels of ribonuclease activity.

PCR and Real-Time PCR, Reverse Transcription

Order Info

Approved for shipment on Wet or Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage Second-Strand Buffer is supplied at a 5X concentration. Store at -20°C.
Quantity 0.5 mL
Shipping Condition Approved for shipment on Wet or Dry Ice
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.